
Latanoprost

Synonym(s):
Isopropyl (5Z,9α,11α,15R)-9,11,15-trihydroxy-17-phenyl-18,19,20-trinor-prost-5-en-1-oate, Isopropyl (Z)-7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-[(3R)-3-hydroxy-5-phenylpentyl]cyclopentyl]hept-5-enoate, Xalatan, Latanoprost
Empirical Formula (Hill Notation):
C26H40O5
CAS Number:
Molecular Weight:
432.59
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-4 of 4