
Inhibitor Resistant Genotyping PCR ReadyMix™, ROX™

Synonym(s):
Inhibitor Resistant Genotyping PCR ReadyMix™
Products
Product Category
Brand
Color
Physical Form
Number of Reactions
Quality Segment
dNTPs
Hot Start
PCR Application
Sample Type
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing