
Hydroxynaphthol blue

Synonym(s):
Hydroxynaphthol blue
Empirical Formula (Hill Notation):
C20H11N2Na3O11S3
CAS Number:
Molecular Weight:
620.47
Products
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing