
Granisetron hydrochloride

Synonym(s):
1-Methyl-N-[(3-endo)-9-methyl-9-azabicyclo[3.3.1]non-3-yl]-1H-indazole-3-carboxamide hydrochloride, Granisetron hydrochloride
Empirical Formula (Hill Notation):
C18H24N4O· HCl
CAS Number:
Molecular Weight:
348.87
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing