
Glycerol carbonate-(carbonyl-13C)

Synonym(s):
4-(Hydroxymethyl)-1,3-dioxolan-2-one-2-13C
Empirical Formula (Hill Notation):
13CC3H6O4
CAS Number:
Molecular Weight:
119.08
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Isotope
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing