
Galantamine hydrobromide

Synonym(s):
Galanthamine hydrobromide, Galantamine hydrobromide
Empirical Formula (Hill Notation):
C17H21NO3 · HBr
CAS Number:
Molecular Weight:
368.27
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing