
Fosinopril Related Compound H

Synonym(s):
Fosinopril related compound H
Empirical Formula (Hill Notation):
C10H15O3P
CAS Number:
Molecular Weight:
214.20
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2