
Fludeoxyglucose

Synonym(s):
2-Fluoro-2-deoxy-D-glucose, 2-Deoxy-2-fluoro-D-glucose, FDG
Empirical Formula (Hill Notation):
C6H11FO5
CAS Number:
Molecular Weight:
182.15
Products
Formula Weight
Manufacturer
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing