
Famotidine Related Compound B

Synonym(s):
N,N′′′-[(1,1-Dioxido-2H-1,2,4,6-thiatriazine-3,5-diyl)bis(2,1-ethanediylthiomethylene-4,2-thiazolediyl)]bisguanidine, 3,5-Bis[2-[[[2-[(diaminomethylene)amino]thiazol-4-yl]methyl]sulphanyl]ethyl]-4H-1,2,4,6-thiatriazine 1,1-dioxide
Empirical Formula (Hill Notation):
C16H23N11O2S5
CAS Number:
Molecular Weight:
561.75
Products
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2