
Etrimfos

Synonym(s):
O-(6-Ethoxy-2-ethyl-4-pyrimidinyl) O,O-dimethyl thiophosphate
Empirical Formula (Hill Notation):
C10H17N2O4PS
CAS Number:
Molecular Weight:
292.29
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing