
Edifenphos

Synonym(s):
O-Ethyl S,S-diphenyl phosphorodithioate
Empirical Formula (Hill Notation):
C14H15O2PS2
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2