
Dimethenamide-P

Synonym(s):
(S)-2-Chloro-N-(2,4-dimethyl-2-thienyl)-N-(2-methoxy-1-methylethyl)acetamide
Empirical Formula (Hill Notation):
C12H18ClNO2S
CAS Number:
Molecular Weight:
275.79
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing