
Dillapiole

Synonym(s):
6-Allyl-4,5-dimethoxy-1,3-benzodioxole
Empirical Formula (Hill Notation):
C12H14O4
CAS Number:
Molecular Weight:
222.24
Products
Brand
Formula Weight
Manufacturer
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing