
Delphinidin 3-galactoside chloride

Synonym(s):
Delphinidin 3-O-β-D-galactoside chloride, Delphinidin 3-O-β-D-galactopyranoside, Delphinidin 3-galactoside chloride, Empetrin chloride
Empirical Formula (Hill Notation):
C21H21ClO12
CAS Number:
Molecular Weight:
500.84
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing