
Daunorubicinone

Synonym(s):
(8S,10S)-8-Acetyl-6,8,10,11-tetrahydroxy-1-methoxy-7,8,9,10-tetrahydrotetracene-5,12-dione, Daunomycinone, Daunorubicinone
Empirical Formula (Hill Notation):
C21H18O8
CAS Number:
Molecular Weight:
398.36
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2