
Clonidine impurity B

Synonym(s):
1-Acetyl-2-[(2,6-dichlorophenyl)amino]-4,5-dihydro-1H-imidazole
Empirical Formula (Hill Notation):
C11H11Cl2N3O
CAS Number:
Molecular Weight:
272.13
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing