
Acetobromo-α-D-glucose

Synonym(s):
1-Bromo-α-D-glucose tetraacetate, 2,3,4,6-Tetra-O-acetyl-α-D-glucopyranosyl bromide
Empirical Formula (Hill Notation):
C14H19BrO9
CAS Number:
Molecular Weight:
411.20
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing