
6-Hydroxynicotinic acid

Synonym(s):
6-Hydroxypyridine-3-carboxylic acid, 2-Hydroxy-5-pyridinecarboxylic acid, 6-Hydroxynicotinic acid
Empirical Formula (Hill Notation):
C6H5NO3
CAS Number:
Molecular Weight:
139.11
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2