
4-Nitrophenyl phosphate disodium salt hexahydrate

Synonym(s):
p-Nitrophenyl phosphate disodium hexahydrate, Disodium 4-nitrophenyl phosphate hexahydrate, di-Sodium 4-nitrophenyl phosphate, pNPP disodium hexahydrate
Linear Formula:
O2NC6H4OP(O)(ONa)2 · 6H2O
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing