
4-Hydroxy-5-methyl-3-furanone

Linear Formula:
C4OH2OCH3OH
Products
Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing