Skip to Content
EE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2-Methoxythiophenol
Synonym(s):
2-Mercaptoanisole, 2-Methoxybenzenethiol, Thioguaiacol
Linear Formula:
CH
3
OC
6
H
4
SH
CAS Number:
7217-59-6
Molecular Weight:
140.20
EC Number:
230-605-7
Products
Product Category
aroma chemicals (1)
building blocks (1)
Brand
Sigma-Aldrich (2)
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Food Grade (1)
Halal (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
184055
2-Methoxythiophenol,
97%
Expand
View Pricing
W415901
2-Methoxythiophenol,
≥97%, FG
Expand
View Pricing