
2-Bromo-6-chloropyrazine

Empirical Formula (Hill Notation):
C4H2BrClN2
CAS Number:
Molecular Weight:
193.43
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing