Skip to Content
Merck
2,2,6-Trimethyl-4H-1,3-dioxin-4-one Arxada quality, ≥93.0% (GC)

2,2,6-Trimethyl-4H-1,3-dioxin-4-one

2,2,6-Trimethyl-4H-1,3-dioxin-4-one Arxada quality, ≥93.0% (GC)
Synonym(s):
Diketene acetone adduct
Empirical Formula (Hill Notation):
C7H10O3
CAS Number:
Molecular Weight:
142.15

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2