
1α,25-Dihydroxyvitamin D3 solution

Empirical Formula (Hill Notation):
C27H44O3
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing