
1,2-Dipalmitoyl-sn-glycerol

Synonym(s):
(S)-1,2-Dipalmitin, (S)-Glycerol 1,2-dipalmitate, 1,2-Dihexadecanoyl-sn-glycerol, DG(16:0/16:0/0:0), Hexadecanoic acid 1,1′-[(1S)-1-(hydroxymethyl)-1,2-ethanediyl] ester, NSC 269964
Empirical Formula (Hill Notation):
C35H68O5
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Tissue
Quality Segment
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing