Skip to Content
Merck
All Photos(1)

Key Documents

48049

Supelco

1,4-Dichlorobenzene-d4 solution

certified reference material, 2000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6D4Cl2
CAS Number:
Molecular Weight:
151.03
UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

2000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/C6H4Cl2/c7-5-1-2-6(8)4-3-5/h1-4H/i1D,2D,3D,4D

InChI key

OCJBOOLMMGQPQU-RHQRLBAQSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,4-Dichlorobenzene-d4 solution has been used as a recovery internal standard in the extraction and analysis of polycyclic aromatic hydrocarbons, polychlorinated biphenyls and organochlorine pesticides in water and sediment samples using gas chromatography method (GC). It has also been used as internal standard in the quantitative determination of polycyclic aromatic hydrocarbons in biomass burning aerosols, using gas chromatography (GC) coupled with desorption electrospray ionization mass spectrometry (DESI-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - Solvent

Flash Point(C)

9.7 °C - Solvent


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct quantification of PAHs in biomass burning aerosols by desorption electrospray ionization mass spectrometry
Li M, et al.
International Journal of Mass Spectrometry, 281, 31-36 (2009)
Occurrence of PAHs, PCBs and organochlorine pesticides in the Tonghui River of Beijing, China
Zhang Z, et al.
Environmental Pollution (Barking, Essex : 1987), 130, 249-261 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service