Skip to Content
Merck
All Photos(1)

Key Documents

NIST2926

Recombinant Human Insulin-like Growth Factor 1 (Frozen)

NIST®SRM®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

form

liquid

packaging

vial of 3 × 0.25 mL each
pkg of 1.005 g

manufacturer/tradename

NIST®

technique(s)

LC/MS: suitable

application(s)

clinical research

storage temp.

−20°C

General description

Recombinant Human Insulin-like Growth Factor 1 (Frozen) is a standard reference material. A unit of SRM 2926 consists of three vials, each containing approximately 0.25 mL of a solution of recombinant human IGF-1.

SRM 2926_cert

SRM 2926_SDS

Application

The Recombinant Human Insulin-like Growth Factor 1 Standard Reference Material (SRM) is primarily intended for use in calibrating mass spectroscopy-based procedures and devices for the determination of insulin-like growth factor 1 (IGF-1) in human serum. It can also be used for the value assignment of control materials.

Features and Benefits

The use of this material will provide confidence in the assignment of values to control materials.

Other Notes

  • SRM 2926 is intended for research use only.
  • Expiration details and storage and use instructions are provided on the NIST certificate
  • Notes on biomaterial handling, disposal, and transport documented in MSDS

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service