Skip to Content
Merck
All Photos(1)

Key Documents

BP227

Metformin hydrochloride

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

1,1-Dimethylbiguanide hydrochloride, Metformin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(=NH)NHC(=NH)N(CH3)2 · HCl
CAS Number:
Molecular Weight:
165.62
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

metformin

form

crystalline

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

mp

223-226 °C (lit.)

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].CN(C)C(=N)NC(N)=N

InChI

1S/C4H11N5.ClH/c1-9(2)4(7)8-3(5)6;/h1-2H3,(H5,5,6,7,8);1H

InChI key

OETHQSJEHLVLGH-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Metformin hydrochloride BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Metformin Tablets
  • Metformin Oral Solution
  • Metformin and Sitagliptin Prolonged-release Tablets
  • Metformin Prolonged-release Tablets
  • Metformin and Sitagliptin Tablets
  • Biochem/physiol Actions

    Metformin is an antidiabetic agent that reduces blood glucose levels and improves insulin sensitivity. Its metabolic effects, including the inhibition of hepatic gluconeogenesis, are mediated at least in part by activation of the LKB1-AMPK (AMP-activated protein kinase) pathway. Activation of this pathway also appears to be involved in the antiproliferative and proapoptotic actions of metformin in cancer cell lines.

    Packaging

    Unit quantity: 100 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.

    Pictograms

    Exclamation mark

    Signal Word

    Warning

    Hazard Statements

    Hazard Classifications

    Acute Tox. 4 Oral - Eye Irrit. 2

    Storage Class Code

    11 - Combustible Solids

    WGK

    WGK 1

    Flash Point(F)

    Not applicable

    Flash Point(C)

    Not applicable


    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Maryam Fatehi-Agdam et al.
    Drug research, 71(1), 17-25 (2020-09-29)
    Gastric cancer is a common gastrointestinal cancer characterized by poor prognosis and chemoresistance. Docetaxel and 5-fluorouracil (5-FU) are frequently used for the treatment of gastric cancer. Despite their potent anti-cancer effects, chemoresistance occurs in metastatic gastric cancer. Metformin, a popular

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service