Skip to Content
Merck
All Photos(3)

Key Documents

229164

Sigma-Aldrich

4-(Phenylazo)diphenylamine

97%

Synonym(s):

4-Anilinoazobenzene, N-Phenyl-4-phenylazoaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5N=NC6H4NHC6H5
CAS Number:
Molecular Weight:
273.33
Beilstein:
749359
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

Assay

97%

form

powder

mp

89-91 °C (lit.)

λmax

411 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

N(c1ccccc1)c2ccc(cc2)\N=N\c3ccccc3

InChI

1S/C18H15N3/c1-3-7-15(8-4-1)19-16-11-13-18(14-12-16)21-20-17-9-5-2-6-10-17/h1-14,19H/b21-20+

InChI key

VXLFYNFOITWQPM-QZQOTICOSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annett Boeddrich et al.
Cell chemical biology, 26(1), 109-120 (2018-11-26)
Self-propagating amyloid-β (Aβ) aggregates or seeds possibly drive pathogenesis of Alzheimer's disease (AD). Small molecules targeting such structures might act therapeutically in vivo. Here, a fluorescence polarization assay was established that enables the detection of compound effects on both seeded and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service