Skip to Content
Merck
All Photos(1)

Key Documents

AB3598

Sigma-Aldrich

Anti-Mitochondria Antibody

serum, Chemicon®

Synonym(s):

Anti-ACNINV3, Anti-FAD, Anti-PS-1, Anti-S182

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

monkey, human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
western blot: suitable

input

sample type neural stem cell(s)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Presenilin 1 is a protein found in brain cells. Presenilin 1 has a major influence on the risk of contracting alzheimer′s disease. Its function is unknown. However, it may be involved in protein production and trafficking, especially during early development. More than 40 different mutations have been found in the presenilin 1 gene, and these mutations associate with early-onset familial alzheimer′s disease. Mutations in presenilin 1 tend to elevate levels of amyloid beta in the blood, cerebrospinal fluid, and brains of those affected by mutations

Specificity

Mitochondria. Recognizes a 62 kD protein of immunoblots of subcellular fractions enriched in mitochondrial fractions.



The antibody shows a moderate titer by ELISA to the immunogen peptide but does not immunoblot any expected presenilin 1 bands. Immunocytochemistry with the serum revealed a mitochondria-like staining that was still present after exhaustive preabsorption with the peptide. The staining co-localizes with CHEMICON′s anti-mitochondria antibody MAB1273.

Immunogen

Presenilin 1 peptide conjugated to KLH.

Application

Anti-Mitochondria Antibody is an antibody against Mitochondria for use in WB & IC.
Immunoblotting: 1:500-1:1000

Immunocytochemistry: 1:100-1:300 on COS7, HeLa, 293 and SK-N-MC cell lines.

Optimal working dilutions must be determined by the end user.
Research Category
Stem Cell Research
Research Sub Category
Neural Stem Cells

Target description

62 kDa

Physical form

Rabbit serum preabsorbed against immunogen peptide. Liquid. Contains 0.01% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze-thaw cycles.

Analysis Note

Control
HeLa whole cell extract, human neuroblastoma SH-SY5Y cells

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher L Yankaskas et al.
Nature biomedical engineering, 3(6), 452-465 (2019-05-08)
The challenge of predicting which patients with breast cancer will develop metastases leads to the overtreatment of patients with benign disease and to the inadequate treatment of aggressive cancers. Here, we report the development and testing of a microfluidic assay
Kang Chi et al.
Cell transplantation, 27(3), 456-470 (2018-05-15)
Parkinson's disease (PD) causes motor dysfunction and dopaminergic cell death. Drug treatments can effectively reduce symptoms but often cause unwanted side effects. Stem cell therapies using cell replacement or indirect beneficial secretomes have recently emerged as potential therapeutic strategies. Although
Wei Wu et al.
Oncotarget, 7(16), 22819-22833 (2016-03-25)
Retinal pigment epithelium (RPE) transplantation is a particularly promising treatment of retinal degenerative diseases affecting RPE-photoreceptor complex. Embryonic stem cells (ESCs) provide an abundant donor source for RPE transplantation. Herein, we studied the time-course characteristics of RPE cells derived from
Yongwoo Yoon et al.
Stem cells (Dayton, Ohio), 38(8), 936-947 (2020-05-07)
Huntington's disease (HD) is a devastating, autosomal-dominant neurodegenerative disease, for which there are currently no disease-modifying therapies. Clinical trials to replace the damaged striatal medium spiny neurons (MSNs) have been attempted in the past two decades but have met with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service