Skip to Content
Merck
All Photos(1)

Key Documents

1.19225

Supelco

Palladium 99+ powdered

for analysis 99+ EMSURE®

Synonym(s):

Palladium 99+ powdered

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pd
CAS Number:
Molecular Weight:
106.42
MDL number:
UNSPSC Code:
12141733
EC Index Number:
231-115-6
NACRES:
NA.21

Quality Level

product line

EMSURE®

Assay

99.0-101.0% Pd basis

impurities

≤10 ppm Al (Aluminium)
≤10 ppm Cd (Cadmium)
≤10 ppm Co (Cobalt)
≤10 ppm Cr (Chromium)
≤10 ppm Mn (Manganese)
≤10 ppm Ni (Nickel)
≤100 ppm Au (Gold)
≤100 ppm Hg (Mercury)
≤100 ppm Ir (Iridium)
≤100 ppm Na (Sodium)
≤100 ppm Os (Osmium)
≤100 ppm Pt (Platinum)
≤100 ppm Rh (Rhodium)
≤100 ppm Ru (Ruthenium)
≤100 ppm Zn (Zinc)
≤50 ppm Ag (Silver)
≤50 ppm Ca (Calcium)
≤50 ppm Cu (Copper)
≤50 ppm Fe (Iron)
≤50 ppm K (Potassium)
≤50 ppm Mg (Magnesium)
≤50 ppm Pb (Lead)

bp

2200-2900 °C

mp

1554 °C

density

12.02 g/cm3 at 20 °C

storage temp.

no temp limit

InChI

1S/Pd

InChI key

KDLHZDBZIXYQEI-UHFFFAOYSA-N

Related Categories

Analysis Note

Identity (elemental analysis): passes test
Assay (Pd): 99.0 - 101.0 %
Ag (Silver): ≤ 50 ppm
Al (Aluminium): ≤ 10 ppm
Au (Gold): ≤ 100 ppm
Ca (Calcium): ≤ 50 ppm
Cd (Cadmium): ≤ 10 ppm
Co (Cobalt): ≤ 10 ppm
Cr (Chromium): ≤ 10 ppm
Cu (Copper): ≤ 50 ppm
Fe (Iron): ≤ 50 ppm
Hg (Mercury): ≤ 100 ppm
Ir (Iridium): ≤ 100 ppm
K (Potassium): ≤ 50 ppm
Mg (Magnesium): ≤ 50 ppm
Mn (Manganese): ≤ 10 ppm
Na (Sodium): ≤ 100 ppm
Ni (Nickel): ≤ 10 ppm
Os (Osmium): ≤ 100 ppm
Pb (Lead): ≤ 50 ppm
Pt (Platinum): ≤ 100 ppm
Rh (Rhodium): ≤ 100 ppm
Ru (Ruthenium): ≤ 100 ppm
Zn (Zinc): ≤ 100 ppm

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service