Skip to Content
Merck
All Photos(2)

Key Documents

1.09632

Supelco

pH-indicator strips pH 5.2 - 7.2 Special indicator

non-bleeding: colorimetric, pH range 5.2-7.2, for pH measurements in meat, graduations and accuracy: 0.4 pH unit, MQuant®

Synonym(s):

pH dip stick pH 5.2 - 7.2 Special indicator, pH test strips pH 5.2 - 7.2 Special indicator

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113034
NACRES:
NB.35

product name

pH-indicator strips pH 5.2 - 7.2 Special indicator, non-bleeding: no, colorimetric, pH range 5.2-7.2, graduations and accuracy accuracy: 0.4 pH unit, MQuant®

product line

MQuant®

Quality Level

feature

non-bleeding: no

graduations and accuracy measuring range

accuracy: 0.4 pH unit

pH range

5.2-7.2

detection method

colorimetric

storage temp.

2-30°C

General description

During the processing of meat, different biochemical processes can lead to a shift in pH from neutral (pH 7) to acidic (5.4-6.7). This can be monitored using this MQuant special indicator test strip designed for the pH measurement directly in meat. Our special method of manufacture gives the pH indicator strips decisive advantages over the indicator paper: since the indicator does not bleed, the strip can be left in the measurement medium without contaminating it. Using these test strips is fairly easy, the pH value is measured semiquantitatively by visual comparison of the two reaction zones on the test strip with the fields of the color scale. The clever Safetyedge boxes allow for safe storage of the test strips. For tests for other parameters that require a specific pH range of the sample (e.g. Spectroquant, see instructions of use), we recommend the use of MQuant pH test strips as pre-test.
for pH measurements in meat non-bleeding pH 5.2 - 5.6 - 6.0 - 6.4 - 6.8 - 7.2 MColorpHast

Legal Information

MCOLORPHAST is a trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service