Skip to Content
Merck
All Photos(1)

Key Documents

1.05565

Supelco

TLC plastic sheets, Cellulose F

pkg of 25 sheets, sheet L × W 20 cm × 20 cm, plastic support

Synonym(s):

Cellulose F Thin-layer Chromatography Plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
NACRES:
NB.21

material

cellulose matrix
plastic support

Quality Level

feature

binder Organic Polymer
fluorescent indicator

packaging

pkg of 25 sheets

technique(s)

thin layer chromatography (TLC): suitable

layer thickness

70-110 μm

sheet L × W

20 cm × 20 cm

storage temp.

2-30°C

General description

25 Plastic sheets 20 x 20 cm

Linkage

Replaces: 5565-7; 5565

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service