Skip to Content
Merck
All Photos(3)

Key Documents

09-310

Sigma-Aldrich

Anti-phospho-EGFR (Tyr1069) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Receptor tyrosine-protein kinase ErbB-1, avian erythroblastic leukemia viral (v-erb-b) oncogene homolog, cell growth inhibiting protein 40, cell proliferation-inducing protein 61, epidermal growth factor receptor, epidermal growth factor receptor (avian

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

primate (based on 100% sequence homology), rat (based on 100% sequence homology), mouse (based on 100% sequence homology), pig (based on 100% sequence homology), horse (based on 100% sequence homology)

technique(s)

ELISA: suitable
immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pTyr1069)

Gene Information

horse ... Egfr(100067755)
human ... EGFR(1956)
mouse ... Egfr(13649)
pig ... Egfr(397070)
rat ... Egf(25313)

General description

EGFR (Epidermal growth factor receptor, ErbB-1) is a receptor tyrosine kinase (RTK) that is one of four members of the EGFR/ErbB family of receptor tyrosine kinases. EGFR plays a key role in the regulation of essential normal cellular processes and in the pathophysiology of hyperproliferative diseases such as cancer. Activation of the EGFR signaling pathway has been linked with increased cell proliferation, angiogenesis, metastasis and decreased apoptosis. Upon ligand binding, EGFR undergoes a transition from an inactive monomeric form to an active dimer. In addition to forming homodimers, EGFR may pair with another member of the ErbB receptor family, such as ErbB2/Her2/neu, to create an activated heterodimer. EGFR dimerization stimulates its intrinsic intracellular protein-tyrosine kinase activity that leads to trans- and auto-phosphorylation of tyrosine residues within the cytoplasmic domain of the receptor tyrosine kinase of five tyrosine (Y) residues on sites Y992, Y1045, Y1068, Y1148 and Y1173 located on the C-terminal domain of EGFR. This autophosphorylation elicits downstream activation of several signal transduction cascades, principally the MAPK, Akt and JNK pathways through Ras, PI3 Kinase, and PLC pathways that ultimately leads to the activation of the various pathways downstream. Tyr1069 is phosphorylated during activation.

Specificity

Reacts strongly and specifically with Tyr1069 phosphorylated EGFR

Immunogen

Epitope: Tyr1069 (cytoplasmic domain)
Tyrosine-phosphorylated epidermal growth factor receptor.

Application

Immunofluroescent Immunocytochemistry:
Research Category
Apoptosis & Cancer

Signaling
Research Sub Category
Kinases & Phosphatases
This Anti-phospho-EGFR (Tyr1069) Antibody is validated for use in ELISA, IC, WB for the detection of phospho-EGFR (Tyr1069).

Quality

Routinely evaluated by Western Blot on untreated and EGF treated A431 lysates.

Western Blot Analysis:
1:1,000 dilution of this antibody detected Tyr1069 phosphorylated EGFR on 10 g of EGF treated A431 lysates.

Target description

~170 kDa

Linkage

Replaces: 04-339

Physical form

Antigen Affinity Purified
Antigen affinity purified rabbit polyclonal in 0.1M Tris-Glycine (pH 7.4) 150mM NaCl, with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8ºC in undiluted aliquots from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution.

Analysis Note

Control
Untreated and EGF treated A431 lysates

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Feng Chen et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 33(7), 1041-1049 (2013-03-28)
Blood-brain barrier (BBB) dysfunction in acute liver failure (ALF) results in increased BBB permeability that often precludes the patients from obtaining a life-saving liver transplantation. It remains controversial whether matrix metalloproteinase-9 (MMP-9) from the injured liver contributes to the deregulation
Immunodetection of the ligand-activated receptor for epidermal growth factor.
Campos-Gonzalez, R and Glenney, J R
Growth Factors, 4, 305-316 (1991)
Yang Yu et al.
Neuroscience, 463, 227-237 (2021-02-05)
Activation of extracellular signal-regulated kinase 1 and 2 (ERK1/2) signaling in cardiovascular regulatory regions of the brain contributes to sympathetic excitation in myocardial infarction (MI)-induced heart failure (HF) by increasing brain renin-angiotensin system (RAS) activity, neuroinflammation, and endoplasmic reticulum (ER)
Alma Y Galvez-Contreras et al.
The European journal of neuroscience, 43(2), 139-147 (2015-09-16)
Phenytoin is a widely used antiepileptic drug that induces cell proliferation in several tissues, such as heart, bone, skin, oral mucosa and neural precursors. Some of these effects are mediated via fibroblast growth factor receptor (FGFR) and epidermal growth factor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service