Skip to Content
Merck
All Photos(1)

Key Documents

330732

Avanti

LightSPLASH

Avanti Research - A Croda Brand

Synonym(s):

LightSPLASH LIPIDOMIX QUANTITATIVE MASS SPEC PRIMARY STANDARD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.28

form

methanol solution

packaging

ampule of 1 × 1 mL (330732-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

SINGLE-VIAL PREPARED LIPIDOMIC ANALYTICAL STANDARD

MIXTURE OF 13 PRIMARY LIPID STANDARDS AT A CONCENTRATION OF 100μG/ML EACH.  EACH SEALED AMPULE CONTAINS 1ML METHANOL SOLUTION. CONCENTRATIONS ARE VERIFIED FOR USE AS A QUANTITATIVE STANDARD.

Mixture Component
  • 15:0-18:1 PC
  • 18:1 Lyso PC 
  • 15:0-18:1 PE
  • 18:1 Lyso PE
  • 15:0-18:1 PG 
  • 15:0-18:1 PI 
  • 15:0-18:1 PS
  • 15:0-18:1-15:0 TG 
  • 15:0-18:1 DG 
  • 18:1 MG 
  • 18:1 Chol ester 
  • d18:1-18:1 SM
  • C15 Ceramide (d18:1/15:0)

Packaging

2ML Amber Glass Sealed Ampule (330732-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
LIPIDOMIX is a trademark of Avanti Polar Lipids, LLC
LightSPLASH is a trademark of Avanti Polar Lipids, LLC

Kit Components Also Available Separately

Product No.
Description
SDS

  • 845875C18:1 Lyso PC, 1-oleoyl-2-hydroxy-sn-glycero-3-phosphocholine, chloroform 100μg/mLSDS

  • 330717C15:0-18:1 PE, Avanti Research - A Croda Brand 100μg/mLSDS

  • 846725C18:1 Lyso PE, 1-oleoyl-2-hydroxy-sn-glycero-3-phosphoethanolamine, chloroform 100μg/mLSDS

  • 330718C15:0-18:1 PG, Avanti Research - A Croda Brand 100μg/mLSDS

  • 330720C15:0-18:1 PI, Avanti Research - A Croda Brand 100μg/mLSDS

  • 330719C15:0-18:1 PS, Avanti Research - A Croda Brand 100μg/mLSDS

  • 330723C15:0-18:1-15:0 TG, Avanti Research - A Croda Brand 100μg/mLSDS

  • 330722C15:0-18:1 DG, Avanti Research - A Croda Brand 100μg/mLSDS

  • 330724C18:1 MG, Avanti Research - A Croda Brand 100μg/mLSDS

  • 700269C18:1 Chol Ester, Avanti Research - A Croda Brand 100μg/mLSDS

  • 860587C18:1 SM (d18:1/18:1(9Z)), Avanti Research - A Croda Brand 860587C 100μg/mLSDS

  • 330730CC15 Ceramide (d18:1/15:0), Avanti Research - A Croda Brand 100μg/mLSDS

See All (12)

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service