Skip to Content
Merck
All Photos(1)

Key Documents

729914

Sigma-Aldrich

Cyclobutanone-2,2,4,4-d4

95 atom % D, 97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4D4H2O
CAS Number:
Molecular Weight:
74.11
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:

isotopic purity

95 atom % D

Assay

97% (CP)

form

liquid

bp

99 °C (lit.)

density

0.989 g/mL at 25 °C (lit.)

mass shift

M+4

storage temp.

2-8°C

SMILES string

[2H]C1([2H])CC([2H])([2H])C1=O

InChI

1S/C4H6O/c5-4-2-1-3-4/h1-3H2/i2D2,3D2

InChI key

SHQSVMDWKBRBGB-RRVWJQJTSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable Liquids

WGK

WGK 3

Flash Point(F)

80.6 °F

Flash Point(C)

27 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service