Skip to Content
Merck
All Photos(1)

Key Documents

336149

Sigma-Aldrich

Rubidium bromide

99.6% trace metals basis

Synonym(s):

Rubidium monobromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
RbBr
CAS Number:
Molecular Weight:
165.37
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.6% trace metals basis

form

powder

impurities

≤4500.0 ppm Trace Metal Analysis

density

3.35 g/mL at 25 °C (lit.)

SMILES string

[Br-].[Rb+]

InChI

1S/BrH.Rb/h1H;/q;+1/p-1

InChI key

JAAGVIUFBAHDMA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service