Merck
Effect of Glucose on GLUT1-Dependent Intracellular Ascorbate Accumulation and Viability of Thyroid Cancer Cells.
Nutrition and cancer (2015-09-19)
Paweł Jóźwiak, Anna Krześlak, Marek Wieczorek, Anna Lipińska
RESUMEN
Enhanced glucose requirement of cancer cells is associated with an increased glucose transport across plasma membrane that is mediated by a family of facilitated glucose transporter proteins, named GLUTs. GLUT1 is the main transporter in thyroid cancer cells. Glucose is the principal physiological substrate of GLUT1; however, it is also capable of transporting of oxidized form of vitamin C [i.e., dehydroascorbic acid (DHAA) which inside the cells is reduced to ascorbic acid (AA)]. The objective of this study was to determine the effect of normo-, hypo-, and hyperglycemia conditions on GLUT1-dependent intracellular ascorbate accumulation and viability of thyroid cancer cells. GLUT1 seems to be the main DHAA transporter in thyroid cancer cells because its knockdown by RNAi reduced DHAA accumulation by more than 80%. The results showed that in thyroid cancer cells high glucose inhibits both transport of AA and DHAA. Inhibition of vitamin C transport by glucose had a cytotoxic effect on the cells. However, stabilization of vitamin C in one of 2 forms (i.e., AA or DHAA) abolished this effect. These results suggest that cytotoxic effect is rather associated with extracellular accumulation of vitamin C and changes of its oxidation state than with intracellular level of ascorbate.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
2-Deoxy-2-[(7-nitro-2,1,3-benzoxadiazol-4-yl)amino]-D-glucose, ≥97% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Atp11c