Merck
Synthesis and SAR of indazole-pyridine based protein kinase B/Akt inhibitors.
Bioorganic & medicinal chemistry (2006-07-18)
Keith W Woods, John P Fischer, Akiyo Claiborne, Tongmei Li, Sheela A Thomas, Gui-Dong Zhu, Robert B Diebold, Xuesong Liu, Yan Shi, Vered Klinghofer, Edward K Han, Ran Guan, Shayna R Magnone, Eric F Johnson, Jennifer J Bouska, Amanda M Olson, Ron de Jong, Tilman Oltersdorf, Yan Luo, Saul H Rosenberg, Vincent L Giranda, Qun Li
RESUMEN
A series of heteroaryl-pyridine containing inhibitors of Akt are reported. The synthesis and structure-activity relationships are discussed, leading to the discovery of a indazole-pyridine analogue (K(i)=0.16 nM). These compounds bind in the ATP binding site, are potent, ATP competitive, and reversible inhibitors of Akt activity. No selectivity amongst the Akt isoforms is observed for this analogue, but there is good selectivity against an panel of other kinases. It is least selective for other members of the AGC family of kinases but is nonetheless 40-fold selective for Akt over PKA. The compound shows cellular activity and significantly slows tumor growth in vivo.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
5-Bromo-2-fluorobenzaldehyde, 97%
Sigma-Aldrich
5-Bromo-2-fluorobenzonitrile, 97%