Merck
Stage-adapted treatment of HIV-associated Hodgkin lymphoma: results of a prospective multicenter study.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology (2012-10-10)
Marcus Hentrich, Marcel Berger, Christoph Wyen, Jan Siehl, Jürgen K Rockstroh, Markus Müller, Gerd Fätkenheuer, Elisabeth Seidel, Maike Nickelsen, Timo Wolf, Ansgar Rieke, Dirk Schürmann, Ralf Schmidmaier, Manfred Planker, Jürgen Alt, Franz Mosthaf, Andreas Engert, Keikawus Arasteh, Christian Hoffmann
RESUMEN
Although the outcome of patients with HIV-related Hodgkin lymphoma (HIV-HL) has markedly improved since the introduction of combined antiretroviral therapy, standard therapy is still poorly defined. This prospective study investigates a stage- and risk-adapted treatment strategy in patients with HIV-HL. Patients with early favorable HIV-HL received two to four cycles of doxorubicin, bleomycin, vinblastine, and dacarbazine (ABVD) followed by 30 Gy of involved-field (IF) radiation. In patients with early unfavorable HIV-HL, four cycles of bleomycin, etoposide, doxorubicin, cyclophosphamide, vincristine, procarbazine, and prednisone (BEACOPP baseline) or four cycles of ABVD + 30 Gy of IF radiation were administered. Six to eight cycles of BEACOPP baseline were given in patients with advanced-stage HIV-HL. In patients with advanced HIV infection, BEACOPP was replaced with ABVD. Of 108 patients (including eight female patients) included in the study, 23 (21%) had early favorable HL, 14 (13%) had early unfavorable HL, and 71 (66%) had advanced-stage HL. The median CD4 count at HL diagnosis was 240/μL. The complete remission rates for patients with early favorable, early unfavorable, and advanced-stage HL were 96%, 100%, and 86%, respectively. The 2-year progression-free survival of the entire study population was 91.7%. Eleven patients (11%) have died, and treatment-related mortality was 5.6%. The 2-year overall survival rate was 90.7% with no significant difference between early favorable (95.7%), early unfavorable (100%), and advanced-stage HL (86.8%). In patients with HIV-HL, stage- and risk-adapted treatment is feasible and effective. The prognosis for patients with HIV-HL may approach that of HIV-negative patients with HL.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Procarbazine hydrochloride, ≥98% (HPLC)