Merck
  • An improved method for simultaneous analysis of aminothiols in human plasma by high-performance liquid chromatography with fluorescence detection.
An improved method for simultaneous analysis of aminothiols in human plasma by high-performance liquid chromatography with fluorescence detection.
Journal of chromatography. A (2010-02-26)
Giorgio Cevasco, Anna Maria Piatek, Carlo Scapolla, Sergio Thea
RESUMEN
Altered levels of aminothiols in biological fluids are thought to be an important risk indicator for several diseases, and reliable methods for the accurate determination of aminothiols concentrations in plasma are thus required. In this paper ammonium 5-bromo-7-fluorobenzo-2-oxa-1,3-diazole-4-sulphonate (SBD-BF) is proposed as a convenient fluorogenic derivatizating reagent for the determination of aminothiols (cysteine, cysteinylglycine, homocysteine and glutathione) by HPLC with fluorescence detection. The reactions of SBD-BF with aminothiols at room temperature are about three-times faster than those of ammonium 7-fluorobenzo-2-oxa-1,3-diazole-4-sulphonate (the most frequently employed reagent) at 60 degrees C. The derivatives of SBD-BF with cysteine, cysteinylglycine, homocysteine and glutathione are easily separated by HPLC and their calibration curves show excellent linearity over the range 0.05-20 micromol/L with excellent r(2) values for all analytes. SBD-BF reacts with thiols under mild conditions, i.e. at 25 degrees C over about 30 min, and is proposed as a suitable fluorogenic reagent for thiol derivatization to be introduced in analytical clinical chemistry. The detection limits of Cys, Cys-Gly, Hcy and GSH at a signal-to-noise ratio of 5 were 0.1 microM for Cys, 0.01 microM for Cys-Gly and Hcy, and 0.02 microM for GSH. Furthermore, validation parameters of the proposed method are quite satisfactory. As an application of this method the determination of thiol derivatives in human plasma was carried out on a number of samples.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Cys-Gly, ≥85% (TLC)
Sigma-Aldrich
4-Bromo-2,6-difluoroaniline, 98%