Merck
Gas-phase acidities of cysteine-polyglycine peptides: the effect of the cysteine position.
Journal of the American Society for Mass Spectrometry (2010-01-29)
Kiran Kumar Morishetti, Betty De Suan Huang, Jessica Marney Yates, Jianhua Ren
RESUMEN
The sequence and conformational effects on the gas-phase acidities of peptides have been studied by using two pairs of isomeric cysteine-polyglycine peptides, CysGly(3,4)NH(2) and Gly(3,4)CysNH(2). The extended Cooks kinetic method was employed to determine the gas-phase acidities using a triple quadrupole mass spectrometer with an electrospray ionization source. The ion activation was achieved via collision-induced dissociation experiments. The deprotonation enthalpies (Delta(acid)H) were determined to be 323.9 +/- 2.5 kcal/mol (CysGly(3)NH(2)), 319.2 +/- 2.3 kcal/mol (CysGly(4)NH(2)), 333.8 +/- 2.1 kcal/mol (Gly(3)CysNH(2)), and 321.9 +/- 2.8 kcal/mol (Gly(4)CysNH(2)), respectively. The corresponding deprotonation entropies (Delta(acid)S) of the peptides were estimated. The gas-phase acidities (Delta(acid)G) were derived to be 318.4 +/- 2.5 kcal/mol (CysGly(3)NH(2)), 314.9 +/- 2.3 kcal/mol (CysGly(4)NH(2)), 327.5 +/- 2.1 kcal/mol (Gly(3)CysNH(2)), and 317.4 +/- 2.8 kcal/mol (Gly(4)CysNH(2)), respectively. Conformations and energetic information of the neutral and anionic peptides were calculated through simulated annealing (Tripos), geometry optimization (AM1), and single point energy calculations (B3LYP/6-31+G(d)), respectively. Both neutral and deprotonated peptides adopt many possible conformations of similar energies. All neutral peptides are mainly random coils. The two C-cysteine anionic peptides, Gly(3,4)(Cys-H)(-)NH(2), are also random coils. The two N-cysteine anionic peptides, (Cys-H)(-)Gly(3,4)NH(2), may exist in both random coils and stretched helices. The two N-cysteine peptides, CysGly(3)NH(2) and CysGly(4)NH(2), are significantly more acidic than the corresponding C-terminal cysteine ones, Gly(3)CysNH(2) and Gly(4)CysNH(2). The stronger acidities of the former may come from the greater stability of the thiolate anion resulting from the interaction with the helix-macrodipole, in addition to the hydrogen bonding interactions.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Polyglycine, mol wt 500-5,000