Merck
  • Probing the effect of intermolecular interaction and understanding the electrostatic moments of anacardic acid in the active site of p300 enzyme via DFT and charge density analysis.
Probing the effect of intermolecular interaction and understanding the electrostatic moments of anacardic acid in the active site of p300 enzyme via DFT and charge density analysis.
Journal of molecular graphics & modelling (2012-02-07)
B Devipriya, P Kumaradhas
RESUMEN
A charge density analysis has been performed on gas phase and docked forms of anacardic acid molecule to understand its charge density distribution, electrostatic moments and the conformation in the active site of p300 enzyme. Here, we report the binding affinity of anacardic acid with the p300 enzyme calculated from docking analysis. The charge density distribution of anacardic acid molecule in the gas phase as well as the docked form has been determined from the high level quantum chemical calculations using HF and DFT methods coupled with AIM theory. The charge density study on both forms of anacardic acid differentiates its structural and the electrostatic properties in different environments. When the molecule enters into the active site of p300 its conformation, charge density distribution, dipole moment and electrostatic potential are significantly altered in comparison to its gas phase structure. In the active site, the molecule adopts different conformations, its pentadecyl chain is found to be highly twisted; the charges are redistributed and the dipole moment increases from 2.37 to 3.17D. Due to the charge redistribution, the electronegative region of carboxyl group increased as it is found small in the gas phase. The comparisons between both forms reveal the flexibility of anacardic acid in the active site.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anacardic acid