Merck
Oxa-Michael Addition to α,β-Unsaturated Nitriles: An Expedient Route to γ-Amino Alcohols and Derivatives.
ChemCatChem (2018-09-29)
Beibei Guo, Douwe S Zijlstra, Johannes G de Vries, Edwin Otten
RESUMEN
Water addition to α,β-unsaturated nitriles would give facile access to the β-hydroxy-nitriles, which in turn can be hydrogenated to the γ-amino alcohols. We have previously shown that alcohols readily add in 1,4-fashion to these substrates using Milstein's Ru(PNN) pincer complex as catalyst. However, attempted water addition to α,β-unsaturated nitriles gave the 3-hydroxynitriles in mediocre yields. On the other hand, addition of benzyl alcohol proceeded in excellent yields for a variety of β-substituted unsaturated nitriles. Subsequent treatment of the benzyl alcohol addition products with TMSCl/FeCl3 resulted in the formation of 3-hydroxy-alkylnitriles. The 3-benzyloxy-alkylnitriles obtained from oxa-Michael addition also could be hydrogenated directly in the presence of acid to give the amino alcohols as their HCl salts in excellent yields. Hydrogenation under neutral conditions gave a mixture of the secondary and tertiary amines. Hydrogenation in the presence of base and Boc-anhydride gave the orthogonally bis-protected amino alcohols, in which the benzyl ether can subsequently be cleaved to yield Boc-protected amino alcohols. Thus, a variety of molecular scaffolds with a 1,3-relationship between O- and N-functional group is accessible starting from oxa-Michael addition of benzyl alcohol to α,β-unsaturated nitriles.