Merck
Sclareol-loaded lipid nanoparticles improved metabolic profile in obese mice.
Life sciences (2019-01-06)
Gabriela Cavazza Cerri, Leandro Ceotto Freitas Lima, Deborah de Farias Lelis, Lucíola da Silva Barcelos, John David Feltenberger, Samuel Vidal Mussi, Renato Sobral Monteiro-Junior, Robson Augusto Souza Dos Santos, Lucas Antônio Miranda Ferreira, Sérgio Henrique Sousa Santos
RESUMEN
Sclareol is a bioactive hydrophobic diterpene in the essential oil isolated from Salvia sclarea (Fam. Lamiaceae). Sclareol has been widely studied due to its anti-inflammatory and antioxidant effects. The present study aimed to evaluate the effects of Sclareol in different formulations (solid lipid nanoparticle and free) on the metabolic profile of obese mice. Swiss male mice were randomly divided into two groups: standard diet (STD) and high-fat diet (HFD). After obesity induction, each group was divided into three treatment groups: free Sclareol (Sc), Sclareol-loaded lipid nanoparticle (L-Sc) and blank lipid nanoparticle (L). Treatments were performed every day during 30 days. L-Sc improves obese mice metabolic profile by decreasing adiposity, ameliorating insulin sensitivity, glucose tolerance and increasing the HDL plasma levels. In addition, L-Sc decreased the expression of NF-KB, MCP-1 and SERBP-1. The use of sclareol together with lipid nanocarriers may be promising for the treatment of metabolic disorders by reducing adipose tissue.