Merck
Tetrahydrogestrinone is a potent androgen and progestin.
The Journal of clinical endocrinology and metabolism (2004-05-06)
Alison K Death, Kristine C Y McGrath, Rymantas Kazlauskas, David J Handelsman
RESUMEN
Tetrahydrogestrinone (THG) was recently identified as a novel steroid used illicitly to improve athletic performance. Although its structure is closely related to gestrinone, a 19-nor progestin, and resembles that of trenbolone, THG was never marketed, so information on its hormonal properties is not known. In this study, we demonstrate that THG is a highly potent androgen and progestin in a yeast-based in vitro bioassay system expressing human androgen and progesterone receptors. It has no estrogenic activity and no antagonism for any of the three steroid receptor classes.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Gestrinone, ≥97% (HPLC)