Documentos
28423-U
Supelco
Dioxin Sample Preparation Kit
About This Item
UNSPSC Code:
13111000
NACRES:
NB.21
agency
suitable for EPA 1613
technique(s)
solid phase extraction (SPE): suitable
application(s)
food and beverages
General description
Kit includes all glassware and connectors (as listed below).
NOTE: Requires, but does not include, multi-layer silica gel columns and dual layer carbon reversible tubes.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Lot/Batch Number
Lo sentimos, en este momento no disponemos de COAs para este producto en línea.
Visite la Librería de documentos
Contenido relacionado
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.