Key Documents
TTRT96P150W-EP
Eppendorf twin.tec® Real-Time PCR Plates 96
Skirted, PCR clean, volume 150 μL, white plate (white wells), pack of 25
About This Item
UNSPSC Code:
41106300
material
polypropylene wells
white plate (white wells)
sterility
non-sterile
feature
skirt
PCR clean
Skirted
packaging
pack of 25
manufacturer/tradename
Eppendorf® 951022015 NA Only
technique(s)
PCR: suitable
size
96 well
volume
150 μL
working volume
150 μL
Categorías relacionadas
Features and Benefits
  • White wells for better reflection
  • High mechanical stability
  • Raised rims for effective sealing
  • Skirted and semi-skirted frames
  • Optimal heat transfer due to reduced wall thickness
  • Autoclavable (121 °C, 20 min)
  • PCR clean and Eppendorf Forensic DNA Grade s available
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
twin.tec is a registered trademark of Eppendorf AG