Key Documents
BUA4817-EP
Buckets for Eppendorf® A-4-81 Rotor
(capacity 7 x 50 mL Falcon(R) centrifuge tubes), pack of 4
About This Item
UNSPSC Code:
41103910
feature
Holds 7 x 50 ml conical tubes
packaging
pack of 4
manufacturer/tradename
Eppendorf® 022638614 (NA)
Eppendorf® 5825730003 (EU)
technique(s)
centrifugation: suitable
capacity
(capacity 7 x 50 mL Falcon(R) centrifuge tubes)
Categorías relacionadas
Legal Information
Eppendorf is a registered trademark of Eppendorf AG