Merck
PHR1350
Supelco
Cytosine
Pharmaceutical Secondary Standard; Certified Reference Material
Sinónimos:
Cytosine, 4-Amino-2-hydroxypyrimidine, 4-Aminopyrimidin-2-(1H)-one
About This Item
Fórmula empírica (notación de Hill):
C4H5N3O
Número de CAS:
Peso molecular:
111.10
Beilstein/REAXYS Number:
2637
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
grade
certified reference material
pharmaceutical secondary standard
Quality Level
agency
traceable to USP 1162148
API family
cytosine
CofA
current certificate can be downloaded
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
mp
>300 °C (lit.)
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-30°C
SMILES string
NC1=NC(=O)NC=C1
InChI
1S/C4H5N3O/c5-3-1-2-6-4(8)7-3/h1-2H,(H3,5,6,7,8)
InChI key
OPTASPLRGRRNAP-UHFFFAOYSA-N
General description
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Application
Cytosine may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by capillary zone electrophoresis and chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.
Analysis Note
These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Other Notes
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Footnote
To see an example of a Certificate of Analysis for this material enter LRAA1102 in the slot below. This is an example certificate only and may not be the lot that you receive.
related product
Referencia del producto
Descripción
Precios
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
Los clientes también vieron
Characterization of alkaloids in Sophora flavescens Ait. by high-performance liquid chromatography-electrospray ionization tandem mass spectrometry
Liu G, et al.
Journal of Pharmaceutical and Biomedical Analysis, 54(5), 1065-1072 (2011)
Development and validation of a procedure for the determination of minoxidil in hair-regrowth formulations using two variants of capillary zone electrophoresis
Patterson SC, et al.
Il Farmaco (Societa Chimica Italiana : 1989), 60(6-7), 547-554 (2005)
Rebecca C Richmond et al.
Human molecular genetics, 24(8), 2201-2217 (2015-01-02)
Maternal smoking during pregnancy has been found to influence newborn DNA methylation in genes involved in fundamental developmental processes. It is pertinent to understand the degree to which the offspring methylome is sensitive to the intensity and duration of prenatal
Gitali Devi et al.
Nucleic acids research, 42(6), 4008-4018 (2014-01-16)
Peptide nucleic acids (PNAs) have been developed for applications in biotechnology and therapeutics. There is great potential in the development of chemically modified PNAs or other triplex-forming ligands that selectively bind to RNA duplexes, but not single-stranded regions, at near-physiological
Benjamin Delatte et al.
Briefings in functional genomics, 12(3), 191-204 (2013-04-30)
Epigenetic genome marking and chromatin regulation are central to establishing tissue-specific gene expression programs, and hence to several biological processes. Until recently, the only known epigenetic mark on DNA in mammals was 5-methylcytosine, established and propagated by DNA methyltransferases and